missing translation for 'onlineSavingsMsg'
Learn More

Neuroligin 1/NLGN1 Antibody, Novus Biologicals™

Product Code. 18334890 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334890 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334890 Supplier Novus Biologicals Supplier No. NBP276348

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Neuroligin 1/NLGN1 Polyclonal specifically detects Neuroligin 1/NLGN1 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Peptide ELISA, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuroligin 1/NLGN1
Applications ELISA, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:4000, Immunofluorescence
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias EC 3.1.1, EC 3.1.1.1, KIAA1070neuroligin-1, MGC45115, neuroligin 1, NL1
Gene Symbols NLGN1
Host Species Goat
Immunogen Peptide with sequence C-TAKQDDPKQQ corresponding to internal region according to NP_055747.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Research Discipline Cognition and Behavior
Primary or Secondary Primary
Gene ID (Entrez) 22871.0
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.