Learn More
Invitrogen™ Neuroligin 1 Monoclonal Antibody (N97A/31), PerCP
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545687
Product Code. 17897982
  • £407.00 / 100µg
Description
Mouse: 99% identity (125/126 amino acids identical). Human: 99% identity (125/126 amino acids identical) >40% identity with Neuroligin-2 and -3. 2 μg/mL of MA5-45687 was sufficient for detection of Neuroligin-1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 120kDa. Does not cross-react with other Neuroligins. This antibody was formerly sold as clone S97A-31.
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neuroligin 1
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Q62765, Q8N2Q7, Q99K10
Nlgn1
Fusion protein amino acids 718-843 (cytoplasmic C-terminus) of rat Neuroligin-1.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry, Western Blot, Immunocytochemistry
N97A/31
PerCP
Nlgn1
6330415N05Rik; BB179718; Kiaa1070; MGC45115; mKIAA1070; neuroligin 1; neuroligin I; Neuroligin-1; NL1; NLG 1; Nlg1; NLGN1
Mouse
Protein G
RUO
116647, 192167, 22871
4°C