Learn More
Invitrogen™ Neuroligin 1 Monoclonal Antibody (N97A/31), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545685
Description
Mouse: 99% identity (125/126 amino acids identical). Human: 99% identity (125/126 amino acids identical) >40% identity with Neuroligin-2 and -3. 2 μg/mL of MA5-45685 was sufficient for detection of Neuroligin-1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 120kDa. Does not cross-react with other Neuroligins. This antibody was formerly sold as clone S97A-31.
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
Specifications
| Neuroligin 1 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Q62765, Q8N2Q7, Q99K10 | |
| Nlgn1 | |
| Fusion protein amino acids 718-843 (cytoplasmic C-terminus) of rat Neuroligin-1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N97A/31 | |
| APC | |
| Nlgn1 | |
| 6330415N05Rik; BB179718; Kiaa1070; MGC45115; mKIAA1070; neuroligin 1; neuroligin I; Neuroligin-1; NL1; NLG 1; Nlg1; NLGN1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 116647, 192167, 22871 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.