missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurokinin A Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA541649
This item is not returnable.
View return policy
Description
The tachykinins belong to an evolutionary conserved family of peptide neurotransmitters that share the c-terminal sequence Phe-X-Gly-Leu-Met-NH2 and have an established role in neurotransmission. The mammalian tachykinins include substance P, neurokinin A (NKA) and neurokinin B (NKB) which exert their effects by binding to specific receptors. Tachykinin peptides are important in the mediation of many physiological and pathological processes including inflammation, pain, migraine headache and allergy induced asthma.Specifications
Neurokinin A | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P20366 | |
TAC1 | |
Recombinant protein Neurokinin A. The antigen corresponds to amino acid range 98-107 of the target protein. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Paraffin), Western Blot | |
C1 | |
Unconjugated | |
TAC1 | |
4930528L02Rik; C-terminal-flanking peptide; Hs.2563; neurokinin 1; neurokinin 2; neurokinin A; neurokinin alpha; neuromedin L; Neuropeptide gamma; neuropeptide K; NK1; NK-1; NK2; Nka; NKNA; NPK; PPT; Ppt5fl; PPTA; PPT-A; PPTA3; preprotachykinin; preprotachykinin A; protachykinin; protachykinin-1; RATPPTA3; SP; Substance K; substance P; TAC; Tac1; TAC2; Tachykinin (substance P, neurokinin A, neuropeptide K, neuropeptide gamma); tachykinin 1; tachykinin 2; tachykinin precursor 1; tachykinin, precursor 1; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma) | |
Mouse | |
Protein A/G | |
RUO | |
6863 | |
-20°C | |
Liquid |