missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-3 Mouse anti-Human, Clone: PCRP-NEUROG3-1E10, Novus Biologicals™
Mouse Monoclonal Antibody
£200.00 - £396.00
Specifications
| Antigen | Neurogenin-3 |
|---|---|
| Clone | PCRP-NEUROG3-1E10 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
| Applications | Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18360606
|
Novus Biologicals
NBP3-13987-20UG |
20 μg |
£200.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18323784
|
Novus Biologicals
NBP3-13987-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurogenin-3 Monoclonal antibody specifically detects Neurogenin-3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
| Neurogenin-3 | |
| 0.2 mg/ml | |
| Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Stem Cells | |
| 10 mM PBS with 0.05% BSA | |
| Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5 | |
| Recombinant human full-lengthNeurogenin-3protein (Uniprot: Q9Y4Z2) | |
| Primary | |
| Store at 4°C. |
| PCRP-NEUROG3-1E10 | |
| Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q9Y4Z2 | |
| 50674 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title