missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Neurogenin-3 Monoclonal antibody specifically detects Neurogenin-3 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | Neurogenin-3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | PCRP-NEUROG3-1E10 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5 |
| Host Species | Mouse |
| Immunogen | Recombinant human full-lengthNeurogenin-3protein (Uniprot: Q9Y4Z2) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?