missing translation for 'onlineSavingsMsg'
Learn More

Neurogenin-3 Antibody (PCRP-NEUROG3-1E10), DyLight 594, Novus Biologicals™

Product Code. 18336672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336672 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336672 Supplier Novus Biologicals Supplier No. NBP314029DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neurogenin-3 Monoclonal antibody specifically detects Neurogenin-3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurogenin-3
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-NEUROG3-1E10
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5
Host Species Mouse
Immunogen Recombinant human full-lengthNeurogenin-3protein (Uniprot: Q9Y4Z2)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 50674
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.