missing translation for 'onlineSavingsMsg'
Learn More

Neurogenin-3 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18424878
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424878 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424878 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP227115JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Neurogenin-3 Polyclonal antibody specifically detects Neurogenin-3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurogenin-3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5
Host Species Rabbit
Immunogen A portion of amino acids 1-69 of mouse Neurog 3 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 50674
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.