missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neurogenin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534472
This item is not returnable.
View return policy
Description
A suggested positive control is mouse small intestine tissue lysate. PA5-34472 can be used with blocking peptide PEP-1514.
Neurogenin-2 is a member of the neurogenin subfamily of basic helix-loop-helix (bHLH) transcription factor genes that play an important role in neurogenesis from migratory neural crest cells. In the course of mouse neurogenesis, Ngn2 and Ngn1 are expressed in distinct progenitor populations residing in the central and peripheral nervous systems. Mutation analyses demonstrate that Ngn1 is required for the determination of neuronal precursors for proximal cranial sensory ganglia and that Ngn2 is necessary for the determination of precursors for epibranchial placode-derived sensory neurons.
Specifications
| Neurogenin 2 | |
| Polyclonal | |
| Unconjugated | |
| NEUROG2 | |
| Ath4a; ATOH4; atonal homolog 4; bHLHa8; Class A basic helix-loop-helix protein 8; helix-loop-helix protein mATH-4A; mATH4A; NEUROG2; neurogenin 2; neurogenin2; neurogenin-2; NGN2; NGN-2; Protein atonal homolog 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11924, 295475, 63973 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P70447, Q9H2A3 | |
| NEUROG2 | |
| A 19 amino acid peptide near the carboxy terminus of human NGN2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction