missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-2 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41195AF594
This item is not returnable.
View return policy
Description
Neurogenin-2 Polyclonal antibody specifically detects Neurogenin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Neurogenin-2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 594 | |
| Atoh4, BHLHA8, bHLHa8MGC46562, Class A basic helix-loop-helix protein 8, Math4A, neurogenin 2, neurogenin-2, ngn-2, NGN2, Protein atonal homolog 4 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human NGN2. The immunogen is located within the last 50 amino acids of NGN2. Amino Acid Squence: CAGSDMDYWQPPPPDKHRYA | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 63973 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction