missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neurogenin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5102697
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Antibody detects endogenous levels of total NEUROG1.
Neurogenin 1 is a basic helix-loop-helix (bHLH) proteins are transcription factors involved in determining cell type during development. NeuroG1 is a bHLH protein with dual cell-fate specification roles. It functions during neurogenesis, and it has also been shown to inhibit the differentiation of neural stem cells into astrocytes. NeuroG1 promotes neurogenesis by functioning as a transcriptional activator, yet it inhibits astrocyte differentiation by compartmentalizing the CREB-binding protein transcription complex away from astrocyte differentiation genes and by inhibiting STAT transcription factors essential for gliogenesis.
Especificaciones
| Neurogenin 1 | |
| Polyclonal | |
| Unconjugated | |
| NEUROG1 | |
| AKA; ATH4C; basic helix-loop-helix transcription factor; bHLHa6; cb260; chunp6899; class A basic helix-loop-helix protein 6; Helix-loop-helix protein mATH-4C; Math4C; Neurod3; NEUROG1; Neurogenic basic-helix-loop-helix protein; neurogenic differentiation 3; neurogenic differentiation factor 3; neurogenin; neurogenin 1; neurogenin related protein-1; neurogenin-1; Neurogenin-related protein 1; NGN; Ngn1; NGN-1; ngr1; transcription factor; Zath4c; zNgn1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18014, 29410, 4762 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70595, P70660, Q92886 | |
| NEUROG1 | |
| A synthesized peptide derived from human NEUROG1(Accession Q92886), corresponding to amino acid residues A137-G187. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido