missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neurogenin 1 Monoclonal Antibody (OTI8G8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524912
This item is not returnable.
View return policy
Description
Neurogenin 1 Monoclonal Antibody for Western Blot, ICC/IF, IP
Neurogenin 1 is a basic helix-loop-helix (bHLH) proteins are transcription factors involved in determining cell type during development. NeuroG1 is a bHLH protein with dual cell-fate specification roles. It functions during neurogenesis, and it has also been shown to inhibit the differentiation of neural stem cells into astrocytes. NeuroG1 promotes neurogenesis by functioning as a transcriptional activator, yet it inhibits astrocyte differentiation by compartmentalizing the CREB-binding protein transcription complex away from astrocyte differentiation genes and by inhibiting STAT transcription factors essential for gliogenesis.
Specifications
| Neurogenin 1 | |
| Monoclonal | |
| 1.00 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92886 | |
| NEUROG1 | |
| Full length human recombinant protein of NEUROG1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Canine | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| OTI8G8 | |
| Unconjugated | |
| NEUROG1 | |
| AKA; ATH4C; basic helix-loop-helix transcription factor; bHLHa6; cb260; chunp6899; class A basic helix-loop-helix protein 6; Helix-loop-helix protein mATH-4C; Math4C; Neurod3; NEUROG1; Neurogenic basic-helix-loop-helix protein; neurogenic differentiation 3; neurogenic differentiation factor 3; neurogenin; neurogenin 1; neurogenin related protein-1; neurogenin-1; Neurogenin-related protein 1; NGN; Ngn1; NGN-1; ngr1; transcription factor; Zath4c; zNgn1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4762, 481516 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction