missing translation for 'onlineSavingsMsg'
Learn More

Neurogenin-1 Antibody (OTI3F9), DyLight 594, Novus Biologicals™

Product Code. 18420404 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420404 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420404 Supplier Novus Biologicals Supplier No. NBP272948DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neurogenin-1 Monoclonal antibody specifically detects Neurogenin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurogenin-1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone OTI3F9
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, NGN, NGN1
Host Species Mouse
Immunogen Full length recombinant protein of human Neurogenin-1 (NP_006152) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4762
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.