missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-1 Antibody (OTI3F9), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72948AF594
This item is not returnable.
View return policy
Description
Neurogenin-1 Monoclonal specifically detects Neurogenin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Neurogenin-1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NEUROG1 | |
| Full length recombinant protein of human Neurogenin-1 (NP_006152) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3F9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, NGN, NGN1 | |
| Mouse | |
| Affinity Purified | |
| Neuroscience | |
| 4762.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction