missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NeuroG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581226
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Neurogenin-3/NEUROG3.
Neurog 3 protein contains one basic helix-loop-helix (bHLH) domain.It is both necessary and sufficient to induce endocrine islet cell differentiation from embryonic pancreatic progenitors. Not only does itinitiates endocrine cell differentiation, italso promotes islet cell maturation and maintains islet function. It is reported that Neurogenin 3 can redirect the differentiation of bipotential secretory progenitors to endocrine rather than goblet cell fate.
Specifications
| NeuroG3 | |
| Polyclonal | |
| Unconjugated | |
| NEUROG3 | |
| ATH4B; Atoh5; atonal homolog 5; BHLHA7; Class A basic helix-loop-helix protein 7; helix-loop-helix protein mATH-4B; LOW QUALITY PROTEIN: neurogenin-3; Math4B; Neurog3; neurogenin 3; Neurogenin 3 (Atonal protein homolog 5) (Helix-loop-helix protein mATH-4B) (MATH4B); neurogenin3; neurogenin-3; ngn3; NGN-3; Protein atonal homolog 5; Relax; transcriptional regulator, Relax | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 50674 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9Y4Z2 | |
| NEUROG3 | |
| A synthetic peptide corresponding to the center region of the human NEUROG3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction