missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEUROG3, Mouse anti-Human, Clone: 2G9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00050674-M11.100ug
This item is not returnable.
View return policy
Description
Sequence: MTPQPSGAPTVQVTRETERSFPRASEDEVTCPTSAPPSPTRTRGNCAEAEEGGCRGAPRKLRARRGGRSRPKSELALSKQRRSRRKKANDRERNRMHNLN&asterisk;Specifications
| NEUROG3 | |
| Monoclonal | |
| Unconjugated | |
| neurogenin 3 | |
| Atoh5/Math4B/bHLHa7/ngn3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2G9 | |
| In 1x PBS, pH 7.4 | |
| NM_020999 | |
| NEUROG3 | |
| NEUROG3 (NP_066279, 1 a.a. ∼ 101 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 50674 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction