missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurofibromin 1 Antibody (McNFn27a), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-153F
This item is not returnable.
View return policy
Description
Neurofibromin 1 Monoclonal specifically detects Neurofibromin 1 in Human, Mouse, Rat, Guinea Pig samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Neurofibromin 1 | |
| Monoclonal | |
| FITC | |
| P21359 | |
| NF1 | |
| A highly conserved N-terminal peptide, corresponding to amino acids 27-41 of human Neurofibromin 1. [UniProt# P21359] | |
| 0.2 mL | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Cytoskeleton Markers, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
| 4763 | |
| Human, Mouse, Rat, Guinea Pig | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| McNFn27a | |
| ELISA, Immunohistochemistry-Paraffin | |
| DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific for full-length or truncated forms of neurofibromin. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction