missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurofibromin 1 Antibody (CL11773), Novus Biologicals™
Mouse Monoclonal Antibody
£317.00 - £473.00
Specifications
| Antigen | Neurofibromin 1 |
|---|---|
| Clone | CL11773 |
| Dilution | Immunocytochemistry/Immunofluorescence 2-10 ug/ml |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623714
|
Novus Biologicals
NBP3-21202-25ul |
25 μg |
£317.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18636464
|
Novus Biologicals
NBP3-21202-100ul |
100 μg |
£473.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurofibromin 1 Monoclonal antibody specifically detects Neurofibromin 1 in Human samples. It is validated for ImmunofluorescenceSpecifications
| Neurofibromin 1 | |
| Immunocytochemistry/Immunofluorescence 2-10 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS | |
| This antibody was developed against a Synthetic Peptide of P21359. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL11773 | |
| Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
| PBS, pH 7.2, 40% glycerol | |
| 4763 | |
| IgG2b | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title