missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Human Neurodegeneration Panel 2, 4plex

Product Code. 16545701
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 16545701

Brand: Invitrogen™ EPX04015832901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ProcartaPlex Human Neurodegeneration Panel 2 4plex enables neurological research by analyzing 4 proteins targets: apolipoprotein E4, clusterin, complement factor H, and fetuin-A in a single well using Luminex xMAP technology.

The ProcartaPlex Human Neurodegeneration Panel 2 4plex enables neurological research by analyzing 4 proteins targets: apolipoprotein E4, clusterin, complement factor H, and fetuin-A in a single well using Luminex xMAP technology. Simultaneous detection of these targets can help explore diseases such as Amyotrophic lateral sclerosis (ALS), Alzheimer's, or Parkinson's disease. The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup. It is not combinable with simplexes or other panels. It complements the Human Neurodegeneration Panel 1 9plex (Cat. No. EPX090-15836-901) that provides 9 additional protein targets in the same research area.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit our ProcartaPlex Immunoassays page for more information and available products.

Target list [bead region]: Apolipoprotein E4 [65], Clusterin (Apo J) [72], Complement Factor H [38], Fetuin-A [62]

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Quantity 96 Tests
Species Human
Combinability Non-Combinable
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
Format Multiplex Kit
For Use With (Equipment) Luminex™ Instruments
Product Line ProcartaPlex
Sample Type Serum, Plasma, Cerebrospinal Fluid
Sample Volume Serum, Plasma, CSF: 25 μL
Content And Storage

• 2 vials Human Standard Mix Z (lyophilized)
• 1 vial Capture Bead Mix (1X)
• 1 vial Biotinylated Detection Antibody Mix (1X)
• 1 bottle Reading Buffer (1X)
• 1 bottle Streptavidin-PE (1X)
• 1 bottle Universal Assay Buffer (10X)
• 1 bottle Wash Buffer (10X)
• 8-tube strip
• Adhesive film
• Flat-bottom 96-well plate, black

Store at 2°C to 8°C

Shipping Condition Wet Ice
Research Area Neurobiology
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.