missing translation for 'onlineSavingsMsg'
Learn More

NeuroD2 Antibody (PCRP-NEUROD2-1G1), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18363844 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363844 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363844 Supplier Novus Biologicals Supplier No. NBP314015AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NeuroD2 Monoclonal antibody specifically detects NeuroD2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuroD2
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-NEUROD2-1G1
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias BHLHA1, bHLHa1neurogenic differentiation factor 2, Class A basic helix-loop-helix protein 1, MGC26304, NDRFneurogenic basic-helix-loop-helix protein, NeuroD2, NeuroD-related factor, neurogenic differentiation 2
Host Species Mouse
Immunogen Recombinant full-length human NeuroD2 protein (Uniprot: Q15784)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4761
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.