Learn More
NEUROD1 Rabbit anti-Human, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human NEUROD1.
Brand: Abnova PAB29535.100uL
Additional Details : Weight : 0.02000kg
Description
This gene encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus. [provided by RefSeq]
Sequence: ASFPVHPYSYQSPGLPSPPYGTMDSSHVFHVKPPPHAYSAALEPFFESPLTDCTSPSFDGPLSPPLSINGNFSFKHEPSAEFEKNYAFTMHYPAATLAGAQSHGSIFSGTAAPRCEIPIDNIMSFDSHSHHERVMSAQLSpecifications
NEUROD1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant human NEUROD1. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
NEUROD1 | |
Recombinant protein corresponding to human NEUROD1. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
BETA2/BHF-1/NEUROD/bHLHa3 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
4760 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |