Learn More
NEUROD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12081-1-AP-20UL
 View more versions of this product
Product Code. 16859583
  • £135.00 / 20µL
Description
NeuroD1 encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus.
Specifications
Specifications
NEUROD1
Polyclonal
Unconjugated
NEUROD1
BETA2, BHF 1, bHLHa3, NEUROD, NEUROD1, neurogenic differentiation 1
Rabbit
Antigen Affinity Chromatography
RUO
18012, 29458, 4760
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13562, Q60867, Q64289
Neurod1
NEUROD1 Fusion Protein Ag2713
20 μL
Primary
Human, Rat, Mouse