missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies NeuroCult™ Proliferation Kit (Mouse & Rat)

Product Code. 17148281
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
17148281 1 Kit 1 set
1 options
This item is not returnable. View return policy

Product Code. 17148281

Brand: STEMCELL Technologies 05702

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Medium kit for expansion of mouse neural stem and progenitor cells

  • NeuroCult™ Proliferation Kit (Mouse & Rat) is a standardized, serum-free medium and supplement kit for the culture of mouse and rat neural stem and progenitor cells from normal tissues or tumor samples
  • When supplemented with appropriate cytokines, NeuroCult™ Proliferation Kit (Mouse & Rat) is optimized to maintain mouse and rat neural stem cells in culture for extended periods of time without the loss of their self-renewal, proliferation, or differentiation potential
  • NOTE: When preparing Complete NeuroCult™ Proliferation Medium, addition of Human Recombinant EGF (Catalog #78006.1) is required
  • When culturing cells obtained from adult mouse or rat, Human Recombinant bFGF (Catalog #78003.1) and Heparin Solution (Catalog #07980) are also required

Specifications

Cell Type Brain Tumor Stem Cells, Neural Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Expansion, Functional Assay, Spheroid Culture, Toxicity Assay
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.