missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies NeuroCult™ Differentiation Kit (Mouse & Rat)
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- NeuroCult™ Differentiation Kit (Mouse & Rat) is a standardized medium and supplement kit for the differentiation of mouse and rat neural stem and progenitor cells into neurons, astrocytes, and oligodendrocytes
Specifications
Specifications
| Cell Type | Brain Tumor Stem Cells, Neural Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Differentiation, Functional Assay |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction