missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies NeuroCult™ Basal Medium (Mouse & Rat)

Product Code. 17138281
Encompass
Change view
Click to view available options
Quantity:
450 mL
Unit Size:
450mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17138281 450 mL 450mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17138281 Supplier STEMCELL Technologies Supplier No. 05700
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Basal medium for culture of mouse neural stem and progenitor cells

  • NeuroCult™ Basal Medium (Mouse & Rat) is a standardized serum-free basal medium designed to be supplemented with NeuroCult™ Proliferation Supplement (Mouse & Rat; Catalog #05701) and appropriate cytokines for the in vitro culture and expansion of mouse and rat neural stem and progenitor cells
  • It can also be supplemented with NeuroCult™ Differentiation Supplement (Mouse & Rat; Catalog #05703) for the differentiation of mouse and rat neural stem and progenitor cells into neurons, astrocytes, and oligodendrocytes
  • NeuroCult™ Basal Medium (Mouse & Rat) is a component of NeuroCult™ Proliferation Kit (Mouse & Rat; Catalog #05702) and NeuroCult™ Differentiation Kit (Mouse & Rat; Catalog #05704)
  • NOTE: When preparing Complete NeuroCult™ Proliferation Medium, addition of Human Recombinant EGF (Catalog #78006.1) is required
  • When culturing cells obtained from adult mouse or rat, Human Recombinant bFGF (Catalog #78003.1) and Heparin Solution (Catalog #07980) are also required

Specifications

Cell Type Brain Tumor Stem Cells, Neural Stem and Progenitor Cells
For Use With (Application) Cell Culture; Colony Assay; Differentiation; Expansion; Functional Assay; Spheroid Culture
Quantity 450 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.