missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurochondrin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13187-1-AP-150UL
This item is not returnable.
View return policy
Description
Probably involved in signal transduction, in the nervous system, via increasing cell surface localization of GRM5 and positively regulating its signaling. Required for the spatial learning process. Acts as a negative regulator of Ca(2+)-calmodulin-dependent protein kinase 2 (CaMK2) phosphorylation. May play a role in modulating melanin-concentrating hormone-mediated functions via its interaction with MCHR1 that interferes with G protein-coupled signal transduction. May be involved in bone metabolism. May also be involved in neurite outgrowth.Specifications
| Neurochondrin | |
| Polyclonal | |
| Unconjugated | |
| NCDN | |
| KIAA0607, NCDN, Neurochondrin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23154, 26562, 89791 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35095, Q9UBB6, Q9Z0E0 | |
| Ncdn | |
| Neurochondrin Fusion Protein Ag3859 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction