missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NEURL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15553891
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15553891 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15553891 Supplier Invitrogen™ Supplier No. PA536495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 62 kDa. Purity is >95% by SDS-PAGE.

NEURL is involved in the determination of cell fate in the neurogenic region of the embryo and plays a role in the determination of cell fate in the central nervous system. NEURL may act as a tumor suppressor whose inactivation could be associated with malignant progression of astrocytic tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEURL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NEURL1
Gene Accession No. O76050
Gene Alias bA416N2.1; E3 ubiquitin-protein ligase NEURL1; h neu; h neuralized 1; h-neu; h-neuralized 1; neu; neu-1; NEUR1; neuralized E3 ubiquitin protein ligase 1; neuralized homolog; neuralized homolog 1A; neuralized-like homolog; neuralized-like protein 1A; NEURL; Neurl1; NEURL1A; RING finger protein 67; RING-type E3 ubiquitin transferase NEURL1; RNF67
Gene Symbols NEURL1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 221-270 of human Neuralized-1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309459, 9148
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.