missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEURL Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26077V
This item is not returnable.
View return policy
Description
NEURL Polyclonal antibody specifically detects NEURL in Human samples. It is validated for Western Blot
Specifications
| NEURL | |
| Polyclonal | |
| Western Blot | |
| h-neu, h-neuralized 1, neu, neu-1, neuralized (Drosophila)-like, neuralized homolog (Drosophila), neuralized-like protein 1A, NEURL1A, NEURL1neuralized-like (Drosophila), RING finger protein 67, RNF67bA416N2.1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NEURL | |
| 0.1 mL | |
| Neuroscience | |
| 9148 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction