missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neuritin Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145395
This item is not returnable.
View return policy
Description
Neuritin belongs to the neuritin family and is expressed in postmitotic-differentiating neurons of the developmental nervous system as well as neuronal structures associated with plasticity in adults. Its expression can be induced by neural activity and neurotrophins. The protein contains a consensus cleavage signal found in glycosylphoshatidylinositol (GPI)-anchored proteins and promotes neurite outgrowth and arborization, indicating its role in promoting neuritogenesis. Overexpression of the protein may be associated with astrocytoma progression. Alternative splicing leads to multiple transcript variants. NRN1 is associated with Severe Congenital Neutropenia 8 and Spinal Muscular Atrophy, Type III.
Specifications
| Neuritin | |
| Polyclonal | |
| Unconjugated | |
| NRN1 | |
| 0710008J23Rik; Candidate plasticity gene 15 protein; candidate plasticity-related gene 15; Cpg15; dJ380B8.2; Neuritin; neuritin 1; NRN; NRN1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100155948, 51299 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q9NPD7 | |
| NRN1 | |
| Recombinant Human NRN1 protein, Ala28-Phe142 (Accession #Q9NPD7). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction