missing translation for 'onlineSavingsMsg'
Learn More

Neurexophilin-3 Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30733697 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733697 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733697 Supplier Novus Biologicals Supplier No. AF5098MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Neurexophilin-3 Polyclonal antibody specifically detects Neurexophilin-3 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurexophilin-3
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias KIAA1159, neurexophilin 3, NPH3neurexophilin-3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Neurexophilin-3/NXPH-3, Gln23-Gly252, Accession # AAI66603
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 11248
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.