missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Neurexophilin-3 Polyclonal antibody specifically detects Neurexophilin-3 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Neurexophilin-3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | KIAA1159, neurexophilin 3, NPH3neurexophilin-3 |
| Host Species | Goat |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant rat Neurexophilin-3/NXPH-3, Gln23-Gly252, Accession # AAI66603 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?