missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurexophilin-1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98356AF647
This item is not returnable.
View return policy
Description
Neurexophilin-1 Polyclonal specifically detects Neurexophilin-1 in Mouse samples. It is validated for ELISA.
Specifications
| Neurexophilin-1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| Alexa Fluor 647 | |
| neurexophilin 1, neurexophilin-1, NPH1 | |
| Produced in rabbits immunized with purified, recombinant Neurexophilin-1 (Q61200; Ala22-Gly271). | |
| 0.1 mL | |
| Neuroscience | |
| 30010 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction