missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuregulin-3/NRG3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97121-50ul
This item is not returnable.
View return policy
Description
Neuregulin-3/NRG3 Polyclonal antibody specifically detects Neuregulin-3/NRG3 in Human samples. It is validated for ELISA
Specifications
| Neuregulin-3/NRG3 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| HRG3, neuregulin 3, neuregulin 3 variant 1, neuregulin 3 variant 10, neuregulin 3 variant 11, neuregulin 3 variant 12, neuregulin 3 variant 13, neuregulin 3 variant 14, neuregulin 3 variant 3, neuregulin 3 variant 4, neuregulin 3 variant 5, neuregulin 3 variant 6, neuregulin 3 variant 7, neuregulin 3 variant 8, neuregulin 3 variant 9, neuregulin-3-like polypeptide, pro-neuregulin-3, membrane-bound isoform, pro-NRG3 | |
| Produced in rabbits immunized with purified, recombinant Human Neuregulin-3/NRG3 (Accession#: ADK90021.1; Glu282-Pro342) | |
| 50 μg | |
| Cancer, Neuroscience, Signal Transduction | |
| 10718 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction