missing translation for 'onlineSavingsMsg'
Learn More

Neuregulin-2 Antibody (3D2), Novus Biologicals™

Product Code. 18378348 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378348 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378348 Supplier Novus Biologicals Supplier No. H00009542M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neuregulin-2 Monoclonal antibody specifically detects Neuregulin-2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuregulin-2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3D2
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100 to 1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004874
Gene Alias divergent of neuregulin-1, Don-1, DON1, HRG2, neuregulin 2, NTAKneural- and thymus-derived activator for ErbB kinases, pro-neuregulin-2, membrane-bound isoform, Pro-NRG2
Host Species Mouse
Immunogen NRG2 (NP_004874, 116 a.a. ~ 215 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SLKSVQDQAYKAPVVVEGKVQGLVPAGGSSSNSTREPPASGRVALVKVLDKWPLRSGGLQREQVISVGSCVPLERNQRYIFFLEPTEQPLVFKTAFAPLD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9542
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.