missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuregulin-1/NRG1, Mouse anti-Human, Clone: NRG1/2752, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79713-100UG
This item is not returnable.
View return policy
Description
Neuregulin-1/NRG1 Monoclonal specifically detects Neuregulin-1/NRG1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications
| Neuregulin-1/NRG1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
| Q02297, Q02297 | |
| NRG1 | |
| Recombinant fragment of human Neuregulin-1/NRG1 protein (around aa 21-242) (exact sequence is proprietary) | |
| 100 μg | |
| Biologically Active Proteins, Chromatin Research, Glia Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction | |
| 3084 | |
| Store at 4°C. | |
| IgG |
| ELISA, Immunofluorescence, Peptide Array | |
| NRG1/2752 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| ARIA, GGF2, GGFglial growth factor, HGLneu differentiation factor, HRG, HRG1, HRGAneuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha (45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction