missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neural/Glial Antigen 2 (NG2) Monoclonal Antibody (9.2.27), Alexa Fluor™ 488, eBioscience™, Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£182.00 - £393.00

Specifications

Antigen Neural/Glial Antigen 2 (NG2)
Clone 9.2.27
Concentration 0.5 mg/mL
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15312060
View Documents
Invitrogen™
53-6504-80
25 μg
£182.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15322060
GREENER_CHOICE
View Documents
Invitrogen™
53-6504-82
100 μg
£393.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: This 9.2.27 monoclonal antibody reacts with human neural/glial antigen 2, which is also known as melanoma chondroitin sulfate proteoglycan (MCSP). This antigen is composed of a 250 kDa N-linked glycoprotein and a >450 kDa proteoglycan. NG2 is present on the surface of >90% of maligt melanomas in addition to some non-melanomic tumors. Moreover, NG2 is expressed on glioma cells, as well as on developing and adult oligodendrocyte precursor cells. Studies have demonstrated the involvement of this protein in tumor cell proliferation, adhesion, migration, and invasion. NG2 also functions as a coreceptor for alpha4beta1 integrin. Finally, expression of NG2 can be used as a prognostic marker for disorders such as acral letiginous melanoma and infantile acute myeloid leukemia. The 9.2.27 antibody has been reported to suppress melanoma tumor growth. Applications Reported: This 9.2.27 has been reported for use in flow cytometric analysis, immunohistochemical staining of formalin-fixed paraffin embedded tissue (IHC-P), and immunocytochemical staining of fixed cells (ICC). Applications Tested: This 9.2.27 antibody has been tested by immunocytochemistry on methanol-fixed A375 cells at less than or equal to 20 μg/mL and by flow cytometric analysis on A375 cells at less than or equal to 0.125 μg/test. A test is defined as the amount (ug) of antibody that will stain a cell sample in a final volume of 100 μL.

A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells.
Specifications

Specifications

Neural/Glial Antigen 2 (NG2)
0.5 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
4732461B14Rik; AN2; AN2 proteoglycan; AN2/NG2 proteoglycan; chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; CSPG4; HMW MAA; HMW-MAA; HSN tumor-specific antigen; Kiaa4232; MCSP; MCSPG; MEL CSPG; melanoma chondroitin sulfate proteoglycan; Melanoma-associated chondroitin sulfate proteoglycan; MEL-CSPG; membrane-spanning proteoglycan NG2; MSK16; nerve/glial antigen 2; Ng2; Proteoglycan AN2
Cspg4
Primary
4°C, store in dark, DO NOT FREEZE!
Cspg4
9.2.27
Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Alexa Fluor 488
Mouse
Human
Q6UVK1
1464
IgG2a κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.