missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neural/Glial Antigen 2 (NG2) Monoclonal Antibody (7.1), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528549
This item is not returnable.
View return policy
Description
This antibody recognizes an extracellular epitope of NG2, the melanoma-associated chondroitin sulfate proteoglycan 4 of Mw approximately 220-300 kDa. Recommended dilution is for 100 μL of whole blood or 1x10^6 cells in suspension.
A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells.
Specifications
| Neural/Glial Antigen 2 (NG2) | |
| Monoclonal | |
| 0.055 mg/mL | |
| PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
| Q6UVK1 | |
| Cspg4 | |
| Human bone marrow stromal cells infected with SV-40. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 7.1 | |
| PE | |
| Cspg4 | |
| 4732461B14Rik; AN2; AN2 proteoglycan; AN2/NG2 proteoglycan; chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; CSPG4; HMW MAA; HMW-MAA; HSN tumor-specific antigen; Kiaa4232; MCSP; MCSPG; MEL CSPG; melanoma chondroitin sulfate proteoglycan; Melanoma-associated chondroitin sulfate proteoglycan; MEL-CSPG; membrane-spanning proteoglycan NG2; MSK16; nerve/glial antigen 2; Ng2; Proteoglycan AN2 | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| 1464 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction