Learn More
Invitrogen™ NeuN Recombinant Rabbit Monoclonal Antibody (14H6L24)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 702022
Description
This antibody is predicted to react with Monkey, Rat and Mouse. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Fox-3, also known as NEUN, HRNBP3, RBFOX3 (RNA binding protein, fox-1) or Fox-1 homolog C, is a 350 amino acid protein that contains one RRM (RNA recognition motif) domain. Localized to both the nucleus and cytoplasm, Fox-3 is suggested to regulate alternative splicing events. Fox-3 is encoded by a gene located on human chromosome 17, which is comprised over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and LiFraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
Specifications
| NeuN | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| A6NFN3, Q8BIF2 | |
| RBFOX3 | |
| Protein corresponding to Human RBFOX3 (aa 1-312). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, RNA Immunoprecipitation, Immunocytochemistry | |
| 14H6L24 | |
| Unconjugated | |
| RBFOX3 | |
| D11Bwg0517e; FLJ56884; FLJ58356; fox 1 homolog (C. elegans) 3; fox1 homolog C; Fox-1 homolog C; FOX3; FOX-3; FOX3NeuN; hexaribonucleotide binding protein 3; hexaribonucleotide binding protein 3 (HRNBP3); Hexaribonucleotide-binding protein 3; Hrnbp3; NEUN; NeuN antigen; Neuna60; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RBFOX3; RFOX3; RGD1560070; RNA binding fox-1 homolog 3; RNA binding protein; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3; RNA binding protein, fox-1 homolog 3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 146713, 287847, 52897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Responsibly packaged |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.