missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NeuN Monoclonal Antibody (5A4B9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548807
This item is not returnable.
View return policy
Description
NeuN Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA
Fox-3, also known as NEUN, HRNBP3, RBFOX3 (RNA binding protein, fox-1) or Fox-1 homolog C, is a 350 amino acid protein that contains one RRM (RNA recognition motif) domain. Localized to both the nucleus and cytoplasm, Fox-3 is suggested to regulate alternative splicing events. Fox-3 is encoded by a gene located on human chromosome 17, which is comprised over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and LiFraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
Specifications
| NeuN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| A6NFN3, Q8BIF2 | |
| RBFOX3 | |
| Purified recombinant fragment of human RBFOX3 (AA: 1-140) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot | |
| 5A4B9 | |
| Unconjugated | |
| RBFOX3 | |
| D11Bwg0517e; FLJ56884; FLJ58356; fox 1 homolog (C. elegans) 3; fox1 homolog C; Fox-1 homolog C; FOX3; FOX-3; FOX3NeuN; hexaribonucleotide binding protein 3; hexaribonucleotide binding protein 3 (HRNBP3); Hexaribonucleotide-binding protein 3; Hrnbp3; NEUN; NeuN antigen; Neuna60; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RBFOX3; RFOX3; RGD1560070; RNA binding fox-1 homolog 3; RNA binding protein; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3; RNA binding protein, fox-1 homolog 3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 146713, 52897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction