missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neudesin/NENF Mouse anti-Human, Mouse, Clone: 4G9E12, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Neudesin/NENF |
|---|---|
| Clone | 4G9E12 |
| Concentration | 1.2 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16807105
|
Proteintech
60131-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16897095
|
Proteintech
60131-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NENF is a secreted protein that is expressed in neurons but not glial cells of the brain. This protein has neurotrophic activity in primary cultured mouse neurons but not mitogenic activity in primary cultured mouse astrocytes. NENF activated mitogen-activated protein (MAP) and phosphotidylinositol-3 kinase pathways and could be inhibited by pertussis toxin but not receptor tyrosine kinases, suggesting that NENF acts through a Gi/Go-protein-coupled receptor.Specifications
| Neudesin/NENF | |
| 1.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CIR2, NENF, NEUDESIN, SCIRP10, SPUF, SPUF protein | |
| Nenf | |
| IgG2a | |
| Protein A | |
| Antibody |
| 4G9E12 | |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q9CQ45, Q9UMX5 | |
| 29937, 66208 | |
| Neudesin/NENF Fusion Protein Ag8387 | |
| Primary | |
| -20°C | |
| NENF |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title