missing translation for 'onlineSavingsMsg'
Learn More

Neudesin Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30735291 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735291 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735291 Supplier Novus Biologicals Supplier No. AF6714JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Neudesin Polyclonal antibody specifically detects Neudesin in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neudesin
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias Cell immortalization-related protein 2, CIR2cell growth-inhibiting protein 47, NEUDESIN, neuron derived neurotrophic factor, Neuron-derived neurotrophic factor, SCIRP10, SCIRP10-related protein, Secreted protein of unknown function, Spinal cord injury related protein 10, SPUF protein, SPUFneudesin
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Neudesin, Gly32-Phe172, Accession # Q9UMX5
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 29937
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.