missing translation for 'onlineSavingsMsg'
Learn More

Neudesin Antibody (7D10), Novus Biologicals™

Product Code. 18354789 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354789 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354789 Supplier Novus Biologicals Supplier No. H00029937M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neudesin Monoclonal antibody specifically detects Neudesin in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neudesin
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 7D10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_037481
Gene Alias Cell immortalization-related protein 2, CIR2cell growth-inhibiting protein 47, NEUDESIN, neuron derived neurotrophic factor, Neuron-derived neurotrophic factor, SCIRP10, SCIRP10-related protein, Secreted protein of unknown function, Spinal cord injury related protein 10, SPUF protein, SPUFneudesin
Host Species Mouse
Immunogen NENF (NP_037481.1, 1 a.a. ~ 172 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVGPAPRRRLRPLAALALVLALAPGLPTARAGQTPRPAERGPPVRLFTEEELARYGGEEEDQPIYLAVKGVVFDVTSGKEFYGRGAPYNALTGKDSTRGVAKMSLDPADLTHDTTGLTAKELEALDEVFTKVYKAKYPIVGYTARRILNEDGSPNLDFKPEDQPHFDIKDEF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 29937
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.