missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEU4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100024
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NEU4.
The protein encoded by this gene belongs to a family of glycohydrolytic enzymes, which remove terminal sialic acid residues from various sialo derivatives, such as glycoproteins, glycolipids, oligosaccharides, and gangliosides. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifications
| NEU4 | |
| Polyclonal | |
| Unconjugated | |
| NEU4 | |
| 9330166I04; LP5125; N-acetyl-alpha-neuraminidase 4; Neu4; Neuraminidase 4; neuraminidase 4 (sialidase); sialidase 4; sialidase-4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 129807, 241159, 316642 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BZL1, Q8WWR8 | |
| NEU4 | |
| A synthesized peptide derived from human NEU4(Accession Q8WWR8), corresponding to amino acid residues P316-M366. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction