missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEU2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546722
This item is not returnable.
View return policy
Description
Peptide sequence: ANVTRLCQVT STDHGRTWSS PRDLTDAAIG PAYREWSTFA VGPGHCLQLH Sequence homology: Dog: 77%; Human: 100%; Pig: 79%.
This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments.
Specifications
| NEU2 | |
| Polyclonal | |
| Unconjugated | |
| NEU2 | |
| Cytosolic sialidase; N-acetyl-alpha-neuraminidase 2; NEU2; neuraminidase 2; neuraminidase 2 (cytosolic sialidase); SIAL2; sialidase 2 (cytosolic sialidase); sialidase-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4759 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y3R4 | |
| NEU2 | |
| Synthetic peptide directed towards the N-terminal of human NEU2 (aa 119-168). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction