missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEU2 Monoclonal Antibody (OTI4F4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525555
This item is not returnable.
View return policy
Description
NEU2 Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments.
Specifications
| NEU2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y3R4 | |
| NEU2 | |
| Full length human recombinant protein of NEU2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| OTI4F4 | |
| Unconjugated | |
| NEU2 | |
| Cytosolic sialidase; N-acetyl-alpha-neuraminidase 2; NEU2; neuraminidase 2; neuraminidase 2 (cytosolic sialidase); SIAL2; sialidase 2 (cytosolic sialidase); sialidase-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4759 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction