missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEU2 Antibody (OTI4F4), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72944PECY7
This item is not returnable.
View return policy
Description
NEU2 Monoclonal specifically detects NEU2 in Human samples. It is validated for Flow Cytometry.
Specifications
| NEU2 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| NEU2 | |
| Full length recombinant protein of human NEU2 (NP_005374) produced in HEK293T cells. | |
| 0.1 mL | |
| 4759.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI4F4 | |
| Flow Cytometry | |
| Cytosolic sialidase, EC 3.2.1.18, N-acetyl-alpha-neuraminidase 2, neuraminidase 2, SIAL2MGC129579, sialidase 2 (cytosolic sialidase), sialidase-2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction