missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEU2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-21649
This item is not returnable.
View return policy
Description
NEU2 Polyclonal specifically detects NEU2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation.
Specifications
| NEU2 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| Cytosolic sialidase, EC 3.2.1.18, N-acetyl-alpha-neuraminidase 2, neuraminidase 2, SIAL2MGC129579, sialidase 2 (cytosolic sialidase), sialidase-2 | |
| Rabbit | |
| 42.25 kDa | |
| 0.1 mg | |
| Primary | |
| This antibody reacts with human NEU2. Based on sequence we expect this antibody to react with neuraminidase from other sources, although specific reactivity has not been confirmed. Cross-reactivity against Neu1 has not yet been established. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| 0.9 mg/ml | |
| Western Blot 1:500-1:2000, ELISA 1:10000-1:50000, Immunohistochemistry, Immunoprecipitation | |
| Q9Y3R4 | |
| NEU2 | |
| This affinity purified NEU2 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal portion near aa 100-125 of Human NEU2. (Uniprot: Q9Y3R4) | |
| Affinity Purified | |
| RUO | |
| 4759 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction