missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NEU1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA592879

Product Code. 16588281

  • £443.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PRKGRLIVCG HGTLERDGVF CLLSDDHGAS WRYGSGVSGI PYGQPKQEND FNPDECQPYE LPDGSVVINA RNQNNYHCHC RIVLRSYDAC DTLRPRDVTF DPELVDPVVA AGAVVTSSGI VFFSNPAHPE FRVNLTLRWS FSNGTSWRKE TVQLWPGPSG YSSLATLEGS MDGEEQAPQL YVLYEKGRNH YTESISVAKI SVYGTL Positive Samples: Mouse liver, Rat liver; Cellular Location: Cell membrane, Cytoplasmic vesicle, Lumenal side, Lysosome lumen, Lysosome membrane, Peripheral membrane protein.

NEU1 is a lysosomal enzyme, which cleaves terminal sialic acid residues from substrates such as glycoproteins and glycolipids. In the lysosome, this enzyme is part of a heterotrimeric complex together with beta-galactosidase and cathepsin A. Mutations in NEU1 gene can lead to sialidosis.The protein encoded by this gene encodes the lysosomal enzyme, which cleaves terminal sialic acid residues from substrates such as glycoproteins and glycolipids. In the lysosome, this enzyme is part of a heterotrimeric complex together with beta-galactosidase and cathepsin A (the latter also referred to as 'protective protein'). Mutations in this gene can lead to sialidosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NEU1
Polyclonal
Unconjugated
NEU1
AA407268; AA407316; Acetylneuraminyl hydrolase; Aglp; Apl; Bat7; Bat-7; EC 3.2.1.18; exo-alpha-sialidase; FLJ93471; G9; G9 sialidase; HLA-B-associated transcript 7; lysosomal sialidase; Map-2; N-acetyl-alpha-neuraminidase 1; NANH; Neu; NEU1; Neu-1; neuraminidase 1; neuraminidase 1 (lysosomal sialidase); SIAL1; sialidase 1; sialidase 1 (lysosomal sialidase); sialidase-1
Rabbit
Affinity Chromatography
RUO
18010, 24591, 4758
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
1.73 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
O35657, Q99519, Q99PW3
NEU1
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.