missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEU-1/Sialidase-1 Antibody (OTI3H2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44877-100ul
This item is not returnable.
View return policy
Description
NEU-1/Sialidase-1 Monoclonal antibody specifically detects NEU-1/Sialidase-1 in Human samples. It is validated for Western Blot
Specifications
| NEU-1/Sialidase-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3H2 | |
| Western Blot 1:2000 | |
| Acetylneuraminyl hydrolase, EC 3.2.1.18, exo-alpha-sialidase, FLJ93471, G9 sialidase, Lysosomal sialidase, N-acetyl-alpha-neuraminidase 1, NANH, NEU, SIAL1, sialidase 1 (lysosomal sialidase), sialidase-1 | |
| Human recombinant protein fragment corresponding to amino acids 48-315 of human NEU1 (NP_000425)produced in E.coli. | |
| 100 μL | |
| metabolism | |
| 4758 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction