missing translation for 'onlineSavingsMsg'
Learn More

NEU-1/Sialidase-1 Antibody (OTI3C4) [DyLight 594], Novus Biologicals™

Product Code. 30688977 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688977 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688977 Supplier Novus Biologicals Supplier No. NBP344878DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NEU-1/Sialidase-1 Monoclonal antibody specifically detects NEU-1/Sialidase-1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEU-1/Sialidase-1
Applications Western Blot
Classification Monoclonal
Clone OTI3C4
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Acetylneuraminyl hydrolase, EC 3.2.1.18, exo-alpha-sialidase, FLJ93471, G9 sialidase, Lysosomal sialidase, N-acetyl-alpha-neuraminidase 1, NANH, NEU, SIAL1, sialidase 1 (lysosomal sialidase), sialidase-1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 48-315 of human NEU1 (NP_000425)produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4758
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.